1-bromo-4-iodonaphthalene 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Bromo-4-iodonaphthalene

H64825.03 H64825.06
H64825.03EA 71.5 EUR
H64825.03 H64825.06
1-bromo-4-iodonaphthalene 98%
1-bromo-4-iodonaphthalene

Purity: 98% CAS No.: 63279-58-3 Molecular Formula: C10H6BrI Formula Weight: 332.97 Physical Form: Solid Sensitivity: Light Sensitive Melting Point: 84-89° Beilstein Reference: 2044950 MDL No.: MFCD01166242

Formel: C₁₀H₆BrI
Molecular Weight: 332.966 g/mol
Schmelzpunkt: 84…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01166242
CAS-Nummer: 63279-58-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR