4-Nitroanthranilsäure 97+%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-4-nitrobenzoic acid

H64831.14 H64831.22
H64831.14EA 55.9 EUR
H64831.14 H64831.22
4-Nitroanthranilsäure 97+%
4-Nitroanthranilsäure

Purity: 97+% CAS No.: 619-17-0 Molecular Formula: C7H6N2O4 Formula Weight: 182.13 Physical Form: Solid Melting Point: 257° lit. Beilstein Reference: 785071 MDL No.: MFCD00007262

Formel: C₇H₆N₂O₄
Molecular Weight: 182,14 g/mol
Schmelzpunkt: 257 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007262
CAS-Nummer: 619-17-0
EINECS: 210-583-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR