4-(Trifluormethyl)piperidin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Trifluoromethylpiperidine
H64840.03
H64840.06
H64840.03EA
173
EUR
H64840.03
H64840.06
4-(Trifluormethyl)piperidin 97%
4-(Trifluormethyl)piperidin
Purity: 97%
CAS No.: 657-36-3
Molecular Formula: C6H10F3N
Formula Weight: 153.15
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00102068
Formel:
C₆H₁₀F₃N Molecular Weight: 153,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00102068 CAS-Nummer: 657-36-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...