5-Chlor-2-pyridincarbonitril 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2-cyanopyridine
H64846.03
H64846.06
H64846.03EA
45.8
EUR
H64846.03
H64846.06
5-Chlor-2-pyridincarbonitril 96%
5-Chloropicolinonitrile
Purity: 96%
CAS No.: 89809-64-3
Molecular Formula: C6H3ClN2
Formula Weight: 138.55
Physical Form: Solid
Appearance: Off-white
Melting Point: 106-108°
Beilstein Reference: 113764
UN No.: UN3439
MDL No.: MFCD03788835
Formel:
C6H3ClN2 Molecular Weight: 138,55 g/mol Schmelzpunkt: 106…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788835 CAS-Nummer: 89809-64-3 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...