3-Pyridazinamin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Pyridazin-3-amine

H64859.03 H64859.06 H64859.14
H64859.03EA 39.9 EUR
H64859.03 H64859.06 H64859.14
3-Pyridazinamin 97%
3-Aminopyridazine

Purity: 97% CAS No.: 5469-70-5 Molecular Formula: C4H5N3 Formula Weight: 95.10 Physical Form: Solid Appearance: White to pale yellow Melting Point: 168-172° Beilstein Reference: 107024 MDL No.: MFCD01529869

Formel: C4H5N3
Molecular Weight: 95,10 g/mol
Schmelzpunkt: 168…172 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01529869
CAS-Nummer: 5469-70-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR