3-Amino-2-pyridincarbonsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Amino-2-pyridinecarboxylic acid, 3-Aminopyridine-2-carboxylic acid, 2-Pyridinecarboxylic acid, 3-amino-

H64861.06 H64861.14 H64861.22
H64861.06EA 54.7 EUR
H64861.06 H64861.14 H64861.22
3-Amino-2-pyridincarbonsäure 97%
3-Aminopicolinic acid

Purity: 98% CAS No.: 1462-86-8 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Brown Melting Point: 218-222° Beilstein Reference: 3600 MDL No.: MFCD00090153

Formel: C₆H₆N₂O₂
Molecular Weight: 138,126 g/mol
Schmelzpunkt: 218…222 °C
MDL: MFCD00090153
CAS-Nummer: 1462-86-8
EINECS: 215-971-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR