2-Bromo-4'-methylpropiophenone 98%
Lieferant: Thermo Scientific
|
Achtung
|
H64881.03
H64881.06
H64881.14
H64881.03EA
33.8
EUR
H64881.03
H64881.06
H64881.14
2-Bromo-4'-methylpropiophenone 98%
2-Bromo-4'-methylpropiophenone
Purity: 98%
CAS No.: 1451-82-7
Molecular Formula: C10H11BrO
Formula Weight: 227.10
Physical Form: Solid
Beilstein Reference: 2207628
MDL No.: MFCD11131402
Formel:
C₁₀H₁₁BrO Lagertemperatur: Raumtemperatur |
MDL:
MFCD11131402 CAS-Nummer: 1451-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...