4-(N,N-Diphenylamino)benzaldehyd 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Diphenylaminobenzaldehyde

H64887.03 H64887.06 H64887.14
H64887.03EA 37.1 EUR
H64887.03 H64887.06 H64887.14
4-(N,N-Diphenylamino)benzaldehyd 98%
4-(N,N-Diphenylamino)benzaldehyd

Purity: 98% CAS No.: 4181-05-9 Molecular Formula: C19H15NO Formula Weight: 273.33 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Pale yellow Odour: Odourless Melting Point: 129-133° lit. Beilstein Reference: 2732795 MDL No.: MFCD00145131

Formel: C₁₉H₁₅NO
Molecular Weight: 273.33 g/mol
Schmelzpunkt: 129…133 °C (Lit.)
MDL: MFCD00145131
CAS-Nummer: 4181-05-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR