2-(4-Nitrophenyl)ethanaminhydrochlorid 98+%
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: 2-(4-Nitrophenyl)ethylamine hydrochloride
H64894.03 H64894.06 H64894.14
H64894.03EA 40.2 EUR
H64894.03 H64894.06 H64894.14
2-(4-Nitrophenyl)ethanaminhydrochlorid 98+%
4-Nitrophenylethylamine hydrochloride
Purity: 98+% CAS No.: 29968-78-3 Molecular Formula: C8H10N2O2.HCl Formula Weight: 202.64 Physical Form: Crystalline powder Appearance: Yellow to yellow-green Melting Point: 200° lit. Beilstein Reference: 3568915 MDL No.: MFCD00012900
Formel: C₈H₁₁ClN₂O₂
Molecular Weight: 202.64 g/mol
Schmelzpunkt: 200 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012900
CAS-Nummer: 29968-78-3
EINECS: 249-980-3