3-(9H-Carbazol-9-yl)phenylboronic acid 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-(9-Carbazolyl)benzeneboronic acid, 3-(9H-Carbazol-9-yl)benzeneboronic Acid, (3-(9H-Carbazol-9-yl)phenyl)boronic acid

H64898.03 H64898.06
H64898.03EA 56 EUR
H64898.03 H64898.06
3-(9H-Carbazol-9-yl)phenylboronic acid 98%
3-(9H-Carbazol-9-yl)phenylboronic acid

Purity: 98% CAS No.: 864377-33-3 Molecular Formula: C18H14BNO2 Formula Weight: 287.13 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Off-white Beilstein Reference: 15530380 MDL No.: MFCD18072506

Formel: C₁₈H₁₄BNO₂
Molecular Weight: 287,13 g/mol
MDL: MFCD18072506
CAS-Nummer: 864377-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR