dibenzo[b,d]furan-2-carbaldehyde ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
dibenzo[b,d]furan-2-carbaldehyde, Dibenzofuran-2-carboxaldehyde
H64905.03
H64905.06
H64905.03EA
97.8
EUR
H64905.03
H64905.06
dibenzo[b,d]furan-2-carbaldehyde ≥97%
dibenzo[b,d]furan-2-carbaldehyde
Purity: 98%
CAS No.: 5397-82-0
Molecular Formula: C13H8O2
Formula Weight: 196.20
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: Off-white
Melting Point: 74°
Beilstein Reference: 141044
MDL No.: MFCD00136272
Formel:
C₁₃H₈O₂ Molecular Weight: 196,205 g/mol Schmelzpunkt: 74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00136272 CAS-Nummer: 5397-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...