10-Phenyl-9-anthraceneboronic acid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (10-Phenylanthracen-9-yl)boronic acid

H64911.03 H64911.06
H64911.03EA 101 EUR
H64911.03 H64911.06
10-Phenyl-9-anthraceneboronic acid 98%
10-Phenyl-9-anthraceneboronic acid

Purity: 98% CAS No.: 334658-75-2 Molecular Formula: C20H15BO2 Formula Weight: 298.15 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Light Sensitive Beilstein Reference: 11434760 MDL No.: MFCD11111989

Formel: C₂₀H₁₅BO₂
Molecular Weight: 298.15 g/mol
MDL: MFCD11111989
CAS-Nummer: 334658-75-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR