tert-Butyl (4-aminocyclohexyl)carbamate 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
N-Boc-1,4-diaminocyclohexane
H64932.03
H64932.06
H64932.03EA
122
EUR
H64932.03
H64932.06
tert-Butyl (4-aminocyclohexyl)carbamate 95%
tert-Butyl (4-aminocyclohexyl)carbamate
Purity: 95%
CAS No.: 195314-59-1
Molecular Formula: C11H22N2O2
Formula Weight: 214.31
Physical Form: Solid
Appearance: White
Beilstein Reference: 13428745
UN No.: UN3259
MDL No.: MFCD01076211
Formel:
C₁₁H₂₂N₂O₂ Molecular Weight: 214,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01076211 CAS-Nummer: 195314-59-1 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...