5-(Trifluormethyl)picolinsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-(Trifluoromethyl)pyridine-2-carboxylic acid

H64937.03 H64937.06 H64937.14
H64937.03EA 43.5 EUR
H64937.03 H64937.06 H64937.14
5-(Trifluormethyl)picolinsäure 95%
5-(Trifluormethyl)picolinsäure

Purity: 95% CAS No.: 80194-69-0 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Physical Form: Powder Appearance: Pale brown Melting Point: 133-137° MDL No.: MFCD04113632

Formel: C₇H₄F₃NO₂
Molecular Weight: 191,11 g/mol
Schmelzpunkt: 133…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04113632
CAS-Nummer: 80194-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR