Dimethyl-2,3-pyridindicarboxylat 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Dimethyl pyridine-2,3-dicarboxylate
H64939.06
H64939.14
H64939.06EA
30.8
EUR
H64939.06
H64939.14
Dimethyl-2,3-pyridindicarboxylat 95%
Dimethyl-2,3-pyridindicarboxylat
Purity: 95%
CAS No.: 605-38-9
Molecular Formula: C9H9NO4
Formula Weight: 195.17
Physical Form: Solid
Melting Point: 53-58°
Beilstein Reference: 159846
MDL No.: MFCD00186549
Formel:
C₉H₉NO₄ Molecular Weight: 195,17 g/mol Schmelzpunkt: 53…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00186549 CAS-Nummer: 605-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...