2-Chlor-5-pyrimidinboronsäure 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Chloropyrimidine-5-boronic acid

H64949.03 H64949.06
H64949.03EA 81.2 EUR
H64949.03 H64949.06
2-Chlor-5-pyrimidinboronsäure 96%
2-Chloro-5-pyrimidineboronic acid

Purity: 96% CAS No.: 1003845-06-4 Molecular Formula: C4H4BClN2O2 Formula Weight: 158.35 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD08063113

Formel: C₄H₄BClN₂O₂
Molecular Weight: 158.35 g/mol
MDL: MFCD08063113
CAS-Nummer: 1003845-06-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR