Ethyl-1-benzylpyrrolidine-3-carboxylate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl 1-benzylpyrrolidine-3-carboxylate
H64962.06
H64962.14
H64962.06EA
200
EUR
H64962.06
H64962.14
Ethyl-1-benzylpyrrolidine-3-carboxylate 97%
Ethyl-1-benzylpyrrolidine-3-carboxylate
Purity: 97%
CAS No.: 5747-92-2
Molecular Formula: C14H19NO2
Formula Weight: 233.31
Physical Form: Liquid
Appearance: Pale yellow
Beilstein Reference: 398388
MDL No.: MFCD04039852
Formel:
C₁₄H₁₉NO₂ Molecular Weight: 233.31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039852 CAS-Nummer: 5747-92-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...