2-Bromo-5-methoxybenzyl bromide 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Bromo-3-(bromomethyl)anisole, 1-Bromo-2-(bromomethyl)-4-methoxybenzene
H64963.06
H64963.14
H64963.06EA
57.1
EUR
H64963.06
H64963.14
2-Bromo-5-methoxybenzyl bromide 97%
2-Bromo-5-methoxybenzyl bromide
Purity: 97%
CAS No.: 19614-12-1
Molecular Formula: C8H8Br2O
Formula Weight: 279.96
Physical Form: Solid
Appearance: Off-white
Melting Point: 90-94° lit.
Beilstein Reference: 1945118
UN No.: UN3261
MDL No.: MFCD00078664
Formel:
C₈H₈Br₂O Molecular Weight: 279,96 g/mol Schmelzpunkt: 90…94 °C (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078664 CAS-Nummer: 19614-12-1 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...