5-Brom-2-nitrophenol 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Brom-3-hydroxy-4-nitrobenzol
H64966.06
H64966.14
H64966.06EA
87.3
EUR
H64966.06
H64966.14
5-Brom-2-nitrophenol 96%
5-Brom-2-nitrophenol
Purity: 96%
CAS No.: 27684-84-0
Molecular Formula: C6H4BrNO3
Formula Weight: 218.01
Physical Form: Crystalline solid
Appearance: Pale yellow
Melting Point: 40-41°
Beilstein Reference: 2329085
MDL No.: MFCD03095027
Formel:
C₆H₄BrNO₃ Schmelzpunkt: 40…41 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095027 CAS-Nummer: 27684-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...