4-Bromimidazol 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Brom-1H-imidazol
H64972.06
H64972.14
H64972.06EA
70
EUR
H64972.06
H64972.14
4-Bromimidazol 98%
4-Bromoimidazole
Purity: 98%
CAS No.: 2302-25-2
Molecular Formula: C3H3BrN2
Formula Weight: 146.97
Physical Form: Solid
Melting Point: 131-135°
Beilstein Reference: 1615
UN No.: UN2811
MDL No.: MFCD00047021
Formel:
C₃H₃BrN₂ Molecular Weight: 146,97 g/mol Schmelzpunkt: 131…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047021 CAS-Nummer: 2302-25-2 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...