Pyrrolidin-3-ol hydrochloride 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(±)-3-Hydroxypyrrolidine hydrochloride
H64975.06
H64975.14
H64975.06EA
49.6
EUR
H64975.06
H64975.14
Pyrrolidin-3-ol hydrochloride 98%
Pyrrolidin-3-ol hydrochloride
Purity: 98%
CAS No.: 86070-82-8
Molecular Formula: C4H9NO.HCl
Formula Weight: 123.58
Physical Form: Solid
Beilstein Reference: 7029051
MDL No.: MFCD03001754
Formel:
C4H10ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001754 CAS-Nummer: 86070-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...