2-Brom-3-nitro-6-picolin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Brom-6-methyl-3-nitropyridin
H64982.06
H64982.14
H64982.06EA
177
EUR
H64982.06
H64982.14
2-Brom-3-nitro-6-picolin 98%
2-Brom-3-nitro-6-picolin
Purity: 98%
CAS No.: 374633-31-5
Molecular Formula: C6H5BrN2O2
Formula Weight: 217.02
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 138200
MDL No.: MFCD03095212
Formel:
C₆H₅BrN₂O₂ Molecular Weight: 217.02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095212 CAS-Nummer: 374633-31-5 UN: 2811 ADR: 6.1 III |