Ethyl-2-(brommethyl)acrylat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 97%
CAS No.: 17435-72-2
Molecular Formula: C6H9BrO2
Formula Weight: 193.04
Storage Temperature: -30°C to -10°C
Flash Point: 81°(178°F)
Physical Form: Liquid
Density: 1.398
Refractive Index: 1.4790
Appearance: Colourless to pale yellow
Boiling Point: 38°/0.8mm
Beilstein Reference: 970108
MDL No.: MFCD00031518
Formel:
C₆H₉BrO₂ Siedepunkt: 38 °C (0,8 mmHg) Dichte: 1,398 Flammpunkt: 81 °C (178 °F) |
MDL:
MFCD00031518 CAS-Nummer: 17435-72-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...