L-Alaninbenzylesterhydrochlorid 98%
Lieferant: Thermo Scientific
Synonyme:
H-Ala-OBzl.HCl, H-Ala-OBzl·HCl
H65699.06
H65699.14
H65699.22
H65699.06EA
45.4
EUR
H65699.06
H65699.14
H65699.22
L-Alaninbenzylesterhydrochlorid 98%
L-Alanine benzyl ester hydrochloride
Purity: 98%
CAS No.: 5557-83-5
Molecular Formula: C10H13NO2.HCl
Formula Weight: 215.68
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00054340
Formel:
C₁₀H₁₃NO₂ Molecular Weight: 215,68 g/mol |
MDL:
MFCD00054340 CAS-Nummer: 5557-83-5 EINECS: 226-920-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...