D-Propargylglycine 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
H-D-Pra-OH, (R)-2-Aminopent-4-ynoic acid, (R)-alpha-Propargylglycine
H65798.03
H65798.06
H65798.03EA
93.2
EUR
H65798.03
H65798.06
D-Propargylglycine 97%
D-Propargylglycine
Purity: 97%
CAS No.: 23235-03-2
Molecular Formula: C5H7NO2
Formula Weight: 113.12
Physical Form: Powder
Appearance: White to off-white
Odour: Odourless
Melting Point: 205° dec.
Optical Rotation: +23° (c=2 in water)
MDL No.: MFCD00237407
Formel:
C₅H₇NO₂ Molecular Weight: 113,116 g/mol Schmelzpunkt: 205 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00237407 CAS-Nummer: 23235-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...