γ-Aminobutyric acid-t-butyl ester hydrochloride 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: tert-Butyl 4-aminobutanoate hydrochloride, H-γ-Abu-OtBu·HCl, tert-Butyl 4-aminobutyrate hydrochloride

H65936.03 H65936.06
H65936.03EA 58.9 EUR
H65936.03 H65936.06
γ-Aminobutyric acid-t-butyl ester hydrochloride 97%
gamma-Aminobutyric acid-t-butyl ester hydrochloride

Purity: 97% CAS No.: 58640-01-0 Molecular Formula: C8H17NO2.HCl Formula Weight: 195.69 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00270592

Formel: C₈H₁₈ClNO₂
Molecular Weight: 195.687 g/mol
MDL: MFCD00270592
CAS-Nummer: 58640-01-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR