(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid 98% (durch HPLC)
Lieferant: Thermo Scientific
Synonyme: (S)-N-FMOC-Octylglycine, N-Fmoc-2-octyl-L-glycine, Fmoc-octyl-Gly-OH
H66031.03 H66031.06
H66031.03EA 110 EUR
H66031.03 H66031.06
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid 98% (durch HPLC)
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid
Purity: 98% CAS No.: 193885-59-5 Molecular Formula: C25H31NO4 Formula Weight: 409.53 Physical Form: Powder Appearance: White Melting Point: 140-143° MDL No.: MFCD01311753
Formel: C₂₅H₃₁NO₄
Molecular Weight: 409,526 g/mol
Schmelzpunkt: 140…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01311753
CAS-Nummer: 193885-59-5