4-Bromresorcin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Bromresorcinol
H66087.06
H66087.14
H66087.06EA
89.5
EUR
H66087.06
H66087.14
4-Bromresorcin 98%
4-Bromresorcin
Purity: 98%
CAS No.: 6626-15-9
Molecular Formula: C6H5BrO2
Formula Weight: 189.01
Physical Form: Powder
Appearance: Pink
Odour: Odourless
Melting Point: 97-100°
Boiling Point: 150°/12mm
Beilstein Reference: 1861280
MDL No.: MFCD00002272
Formel:
C₆H₅BrO₂ Molecular Weight: 189,01 g/mol Siedepunkt: 150 °C (12 mmHg) Schmelzpunkt: 97…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002272 CAS-Nummer: 6626-15-9 EINECS: 229-586-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...