2-Hydroxymethyloxetane 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Oxetanemethanol, Oxetan-2-ylmethanol, 2,4-Epoxybutanol

H66218.MD H66218.03
H66218.MDEA 105 EUR
H66218.MD H66218.03
2-Hydroxymethyloxetane 96%
2-Hydroxymethyloxetane

Purity: 96% CAS No.: 61266-70-4 Molecular Formula: C4H8O2 Formula Weight: 88.11 Storage Temperature: -30°C to -10°C Physical Form: Liquid Density: 1.080 Refractive Index: 1.4460 Sensitivity: Moisture Sensitive Appearance: Colourless to yellow Boiling Point: 98°/15mm MDL No.: MFCD00142933

Formel: C₄H₈O₂
Molecular Weight: 88,11 g/mol
Siedepunkt: 98 °C (15 mmHg)
Dichte: 1,080
Lagertemperatur: Gefrierschrank
MDL: MFCD00142933
CAS-Nummer: 61266-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR