2-(Bromomethyl)oxetane 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H66276.MD
H66276.03
H66276.MDEA
305
EUR
H66276.MD
H66276.03
2-(Bromomethyl)oxetane 96%
2-(Bromomethyl)oxetane
Purity: 96%
CAS No.: 939759-23-6
Molecular Formula: C4H7BrO
Formula Weight: 151.00
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Colourless to pale yellow
MDL No.: MFCD08544405
Formel:
C₄H₇BrO Molecular Weight: 151.003 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08544405 CAS-Nummer: 939759-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...