5-Iodo-2-methoxybenzaldehyde 97%
Lieferant: Thermo Scientific
|
Achtung
|
H66320.03
H66320.06
H66320.03EA
69.1
EUR
H66320.03
H66320.06
5-Iodo-2-methoxybenzaldehyde 97%
5-Iodo-2-methoxybenzaldehyde
Purity: 97%
CAS No.: 42298-41-9
Molecular Formula: C8H7IO2
Formula Weight: 262.04
Physical Form: Solid
Sensitivity: Air and Light Sensitive
Appearance: White to off-white to yel
Melting Point: 140-146°
MDL No.: MFCD04128999
Formel:
C₈H₇IO₂ Schmelzpunkt: 140…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04128999 CAS-Nummer: 42298-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...