Ethyl-6-chloroindole-2-carboxylate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl 6-chloroindole-2-carboxylate
H66483.03
H66483.06
H66483.03EA
64.5
EUR
H66483.03
H66483.06
Ethyl-6-chloroindole-2-carboxylate 97%
Ethyl-6-chloroindole-2-carboxylate
Purity: 97%
CAS No.: 27034-51-1
Molecular Formula: C11H10ClNO2
Formula Weight: 223.66
Physical Form: Solid
Beilstein Reference: 169178
MDL No.: MFCD03084731
Formel:
C₁₁H₁₀ClNO₂ Molecular Weight: 223.66 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03084731 CAS-Nummer: 27034-51-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...