Borane-2-methylpyridine complex 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Picoline Borane, 2-Methylpyridine Borane, Borane - 2-Picoline Complex
H66642.06
H66642.14
H66642.06EA
85.8
EUR
H66642.06
H66642.14
Borane-2-methylpyridine complex 95%
Borane-2-methylpyridine complex
Purity: 95%
CAS No.: 3999-38-0
Molecular Formula: C6H7N.BH3
Formula Weight: 106.96
Storage Temperature: 2°C to 8°C
Flash Point: 100°(212°F)
Physical Form: Solid
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 44-50°
Solubility: Slightly sol. in water, cyclohexane, very sol. in methanol, THF, diglyme, and benzene
UN No.: UN2813
MDL No.: MFCD07784361
Formel:
C₆H₁₀BN Molecular Weight: 106.96 g/mol Schmelzpunkt: 44…50 °C Flammpunkt: 100 °C (212 °F) |
MDL:
MFCD07784361 CAS-Nummer: 3999-38-0 ADR: 4.3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...