Methyl-N-acetyl-L-tryptophanate 95%
Lieferant: Thermo Scientific
Synonyme:
N-Acetyl-L-tryptophan methyl ester, Methyl N-acetyl-L-tryptophanate, Ac-Trp-OMe
H66937.06
H66937.14
H66937.06EA
75.1
EUR
H66937.06
H66937.14
Methyl-N-acetyl-L-tryptophanate 95%
Methyl-N-acetyl-L-tryptophanate
Purity: 95%
CAS No.: 2824-57-9
Molecular Formula: C14H16N2O3
Formula Weight: 260.29
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white to white
MDL No.: MFCD02728656
Formel:
C₁₄H₁₆N₂O₃ Molecular Weight: 260,293 g/mol |
MDL:
MFCD00022759 CAS-Nummer: 2824-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...