5-Bromo-4-chloro-3-indolyl-α-D-galactoside, Pulver
Lieferant: Thermo Scientific
Synonyme:
X-a-Gal, 5-Bromo-4-Chloro-3-Indolyl-a-D Galactopyranoside, X-alpha-Gal
J60151.MD
J60151.03
J60151.MDEA
212
EUR
J60151.MD
J60151.03
5-Bromo-4-chloro-3-indolyl-α-D-galactoside, Pulver
5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside
Substrate for beta-galactosidase
CAS No.: 107021-38-5
Molecular Formula: C14H15BrClNO6
Formula Weight: 408.63
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00063780
Applications: Substrate for beta-galactosidase
Formel:
C₁₄H₁₅BrClNO₆ Molecular Weight: 408,63 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063780 CAS-Nummer: 107021-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...