Nitrazine yellow
Lieferant: Thermo Scientific
Synonyme:
Sodium 3-((2,4-dinitrophenyl)diazenyl)-4-hydroxynaphthalene-2,7-disulphonate
J60281.03
J60281.06
J60281.14
J60281.03EA
26.7
EUR
J60281.03
J60281.06
J60281.14
Nitrazine yellow
Nitrazine yellow
CAS No.: 5423-07-4
Molecular Formula: C16H8N4Na2O11S2
Formula Weight: 542.36
Physical Form: Powder
Appearance: Brown to yellow to orange
Solubility: Dissolve in water at 1mg/ml
MDL No.: MFCD00003938
Applications: pH indicator
Formel:
C₁₆H₈N₄Na₂O₁₁S₂ Molecular Weight: 542,36 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003938 CAS-Nummer: 5423-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...