Potassium oxonate

Lieferant: Thermo Scientific

Synonyme: Potassium Allantoxanate, Oxonic acid Potassium Salt, Potassium 4,6-Dihydroxy-1,3,5-triazine-2-carboxylate, Allantoxanic acid Potassium Salt, Potassium 4,6-dioxo-1,4,5,6-tetrahydro-1,3,5-triazine-2-carboxylate

J60400.06 J60400.14 J60400.22
J60400.06EA 37.9 EUR
J60400.06 J60400.14 J60400.22
Potassium oxonate
Potassium oxonate

CAS No.: 2207-75-2 Molecular Formula: C4H2KN3O4 Formula Weight: 195.18 Physical Form: Powder Appearance: Off-white Melting Point: 300° MDL No.: MFCD00010565


Applications: Inhibitor of uricase

Formel: C₄H₃N₃O₄
Molecular Weight: 195,18 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010565
CAS-Nummer: 2207-75-2
EINECS: 218-627-5
Merck Index: 14,6947

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR