Chlorogenic acid

Lieferant: Thermo Scientific

Synonyme: (1S,3R,4R,5R)-3-(((3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1,4,5-trihydroxycyclohexanecarboxylic acid, 3-O-Caffeoylquinic acid, 3-(3,4-Dihydroxycinnamoyl)quinic acid, (1S,3R,4R,5R)-3-[[3-(3,4-Dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-1,4,5-trihydroxycyclohexanecarboxylic acid

J60457.MD J60457.03 J60457.06
J60457.MDEA 38.6 EUR
J60457.MD J60457.03 J60457.06
Chlorogenic acid
Chlorogenic acid

CAS No.: 327-97-9 Molecular Formula: C16H18O9 Formula Weight: 354.31 Physical Form: Powder Appearance: White/pale yellow/beige Melting Point: 210° dec. Solubility: Soluble in ethanol, DMSO, or DMF MDL No.: MFCD00003862


Applications: An analog of caffeic acid

Formel: C₁₆H₁₈O₉
Molecular Weight: 354,31 g/mol
Schmelzpunkt: 210 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003862
CAS-Nummer: 327-97-9
EINECS: 206-325-6
Merck Index: 14,2142

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR