Vincamine 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Vincamine base, Methyl (41S,12S,13aS)-13a-ethyl-12-hydroxy-2,3,41,5,6,12,13,13a-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate
J60545.03
J60545.06
J60545.03EA
91.3
EUR
J60545.03
J60545.06
Vincamine 98%
Vincamine
Purity: 98%
CAS No.: 1617-90-9
Molecular Formula: C21H26N2O3
Formula Weight: 354.45
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale yellow
Melting Point: 232°
MDL No.: MFCD00078054
Applications: A vasodilator
Formel:
C₂₁H₂₆N₂O₃ Molecular Weight: 354,45 g/mol Schmelzpunkt: 232 °C |
MDL:
MFCD00078054 CAS-Nummer: 1617-90-9 EINECS: 216-576-3 Merck Index: 14,9983 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...