3-Indoxyl phosphate disodium salt
Lieferant: Thermo Scientific
Synonyme:
3-Indolyl phosphate disodium salt
J60552.MD
J60552.03
J60552.MDEA
116
EUR
J60552.MD
J60552.03
3-Indoxyl phosphate disodium salt
3-Indoxyl phosphate disodium salt
CAS No.: 3318-43-2
Molecular Formula: C8H6NNa2O4P
Formula Weight: 257.09
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD00040646
Applications: A histochemical substrate for alkaline phosphatase
Formel:
C₈H₆NNa₂O₄P Molecular Weight: 257,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040646 CAS-Nummer: 3318-43-2 EINECS: 222-014-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...