Prilocaine hydrochloride 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Prilocaine HCl, 2-(Propylamino)-N-(o-tolyl)propanamide hydrochloride, 2-(Propylamino)-N-(o-tolyl)propionamide Hydrochloride
J60648.03
J60648.06
J60648.09
J60648.03EA
76.8
EUR
J60648.03
J60648.06
J60648.09
Prilocaine hydrochloride 98%
Prilocaine hydrochloride
Purity: 98%
CAS No.: 1786-81-8
Molecular Formula: C13H20N2O.HCl
Formula Weight: 256.77
Physical Form: Powder
Appearance: White
Melting Point: 167-168°
MDL No.: MFCD00079279
Applications: A local anesthetic
Formel:
C₁₃H₂₀N₂O Molecular Weight: 256.77 g/mol Schmelzpunkt: 167…168 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079279 CAS-Nummer: 1786-81-8 EINECS: 217-244-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...