Fasudilhydrochlorid ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-(1-Homopiperazinyl)sulphonylisoquinoline Hydrochloride, 1-(5-Isoquinolinesulphonyl)homopiperazine Hydrochloride, Fasudil HCl, 5-((1,4-Diazepan-1-yl)sulphonyl)isoquinoline hydrochloride
J60751.MD
J60751.ME
J60751.03
J60751.MDEA
266
EUR
J60751.MD
J60751.ME
J60751.03
Fasudilhydrochlorid ≥99%
Fasudil hydrochloride
Purity: 99+%
CAS No.: 105628-07-7
Molecular Formula: C14H17N3O2S.HCl
Formula Weight: 327.83
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00943198
Formel:
C₁₄H₁₇N₃O₂S Molecular Weight: 327,83 g/mol |
MDL:
MFCD00943198 CAS-Nummer: 105628-07-7 Merck Index: 14,3942 |