Tamoxifen citrate
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(Z)-1-(4-Dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate
J60955.03
J60955.06
J60955.03EA
193
EUR
J60955.03
J60955.06
Tamoxifen citrate
Tamoxifen citrate
CAS No.: 54965-24-1
Molecular Formula: C26H29NO.C6H8O7
Formula Weight: 563.65
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive; Hygroscopic
Appearance: White to off-white
MDL No.: MFCD00058321
Applications: Inhibitor of protein kinase C and induces apoptosis in cancer cells
Formel:
C₂₆H₂₉NO Molecular Weight: 563,65 g/mol |
MDL:
MFCD00058321 CAS-Nummer: 54965-24-1 EINECS: 259-415-2 Merck Index: 14,9048 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...