IGEPAL® CA-630

Lieferant: Thermo Scientific
Gefahr

Synonyme: Octylphenyl-polyethylene glycol

J61055.AE J61055.AP
J61055.AEEA 89.1 EUR
J61055.AE J61055.AP
IGEPAL® CA-630
IGEPAL® CA-630

Detergent, equivalent to Nonidet P-40. For solubilizing membrane proteins.


IGEPAL® CA-630 is used as a detergent in paper industry, hand cleaners, metal cleaners. It is suitable for solubilization, isolation and purification of membrane protein complexes and in two-dimensional gel electrophoresis.


Applications: Detergent, equivalent to Nonidet P-40. For solubilizing membrane proteins


Bitte beachten IGEPAL® is a registered trademark of Rhodia Operations. Incompatible with oxidizing agents and reducing agents.

Formel: (C₂H₄O)nC₁₄H₂₂O
Dichte: 1,060
Lagertemperatur: Raumtemperatur
MDL: MFCD00132505
CAS-Nummer: 9002-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR