Benzydamine hydrochloride
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzydamine HCl, 1-Benzyl-3-[3-(dimethylamino)propoxy]-1H-indazole Hydrochloride, 3-((1-Benzyl-1H-indazol-3-yl)oxy)-N,N-dimethylpropan-1-amine hydrochloride
CAS No.: 132-69-4
Molecular Formula: C19H23N3O.HCl
Formula Weight: 345.87
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White
Melting Point: 156-161°
Solubility: Soluble in water, ethanol, chloroform, n-butanol or DMSO
MDL No.: MFCD00078957
Applications: Nonsteroidal analgesic and anti-inflammatory compound. A prostaglandin synthase inhibitor
Formel:
C₁₉H₂₃N₃O Molecular Weight: 345,87 g/mol Schmelzpunkt: 156…161 °C |
MDL:
MFCD00078957 CAS-Nummer: 132-69-4 EINECS: 205-076-0 Merck Index: 14,1122 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...