Solvent black 5
Lieferant: Thermo Scientific
Synonyme:
2-Amino-N,N-dipropylacetamide hydrochloride, Nigrosin B, alcohol soluble, Nigrosin soluble in spirit
J61186.14
J61186.22
J61186.A1
J61186.14EA
26.1
EUR
J61186.14
J61186.22
J61186.A1
Solvent black 5
Solvent black 5
CAS No.: 11099-03-9
Physical Form: Powder
Appearance: Black
MDL No.: MFCD00149005
Formel:
C₈H₁₉ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071316 CAS-Nummer: 11099-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...