Mevastatin 98%

Lieferant: Thermo Scientific

Synonyme: (1S,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (S)-2-methylbutanoate, Compactin

J61357.X0 J61357.MA J61357.MB
J61357.X0EA 53.5 EUR
J61357.X0 J61357.MA J61357.MB
Mevastatin 98%
Mevastatin

Inhibitor of HMG-CoA reductase. Induces apoptosis.


Applications: Inhibitor of HMG-CoA reductase. Induces apoptosis

Formel: C₂₃H₃₄O₅
Molecular Weight: 390,52 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05662341
CAS-Nummer: 73573-88-3
Merck Index: 14,6164

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR