Furosemid ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 5-(Aminosulphonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, 4-Chloro-N-furfuryl-5-sulphamoylanthranilic acid

J61457.06 J61457.09 J61457.14
J61457.06EA 92.5 EUR
J61457.06 J61457.09 J61457.14
Furosemid ≥97%
Furosemide

Purity: 97+% CAS No.: 54-31-9 Molecular Formula: C12H11ClN2O5S Formula Weight: 330.74 Physical Form: Crystalline powder Appearance: Off-white Melting Point: 206-209° Solubility: Soluble in acetone, DMF or methanol. Slightly soluble in water MDL No.: MFCD00010549


Applications: Loop diuretic that inhibits the Na+/2Cl-/K+ (NKCC) cotransporter

Formel: C₁₂H₁₁ClN₂O₅S
Molecular Weight: 330.74 g/mol
Schmelzpunkt: 206…209 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010549
CAS-Nummer: 54-31-9
EINECS: 200-203-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR