SB203580 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: SB 203580, 4-(5-(4-Fluorophenyl)-2-(4-(methylsulphinyl)phenyl)-1H-imidazol-4-yl)pyridine

J61482.MB J61482.MF J61482.MC
J61482.MBEA 136 EUR
J61482.MB J61482.MF J61482.MC
SB203580 98%
SB203580

Applications: Highly specific new inhibitor of p38 kinase and MAP Kinase homologues

Formel: C₂₁H₁₆FN₃OS
Molecular Weight: 377,4 g/mol
Schmelzpunkt: 235…247 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00922198
CAS-Nummer: 152121-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR