Cyclopamine 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(2'R,3S,3'R,3a'S,6aS,6bS,6'S,7a'R,11aS,11bR)-3',6',10,11b-tetramethyl-1,2,3,3a',4,4',5',6,6a,6b,6',7,7',7a',8,11,11a,11b-octadecahydro-3'H-spiro[benzo[a]fluorene-9,2'-furo[3,2-b]pyridin]-3-ol
J61528.MB
J61528.MF
J61528.MBEA
245
EUR
J61528.MB
J61528.MF
Cyclopamine 99%
Cyclopamine
Purity: 99+%
CAS No.: 4449-51-8
Molecular Formula: C27H41NO2
Formula Weight: 411.62
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 234-237°
MDL No.: MFCD01735266
Applications: Inhibits hedgehog/smoothened signaling. A steroidal alkaloid
Formel:
C₂₇H₄₁NO₂ Molecular Weight: 411,63 g/mol Schmelzpunkt: 234…237 °C |
MDL:
MFCD09878269 CAS-Nummer: 4449-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...